Learn More
Andwin Scientific LCRY 1400 CRYOBABIES 1X1
SDP
Catalog No. NC0269239
Encompass_Preferred
The Cryo-Babies labels can withstand most organic solvents, caustic agents, temperatures from -196C to 100C. This product can still be utilized while under oven, autoclaving, or extreme freezer temperatures. The labels can also be used on several surfaces like plastics, glass, and metals without cracking, peeling, or degrading. This product is sold in a pack of 1600 white labels measuring 1" L x 1" W, which is a perfect size for use with microscope slides.
Catalog No. NC0269239 Supplier Andwin Scientific Supplier No. LCRY1400
The Cryo-Babies labels can withstand most organic solvents, caustic agents, temperatures from -196C to 100C. This product can still be utilized while under oven, autoclaving, or extreme freezer temperatures. The labels can also be used on several surfaces like plastics, glass, and metals without cracking, peeling, or degrading. This product is sold in a pack of 1600 white labels measuring 1" L x 1" W, which is a perfect size for use with microscope slides.