Learn More
Medchemexpress LLC LCS-1 100mg | 41931-13-9 | 255.10 | C11H8Cl2N2O | 100 MG
SDP
Catalog No. 5000228871
Encompass_Preferred
LCS-1 is a small-molecule inhibitor of superoxide dismutase 1 (SOD1) used in research to induce apoptosis and study cancer cell biology. The compound is provided as a solid powder and characterized by CAS 41931-13-9, molecular formula C11H8Cl2N2O, and molecular weight 255.10 g·mol-1.
  • Inhibits superoxide dismutase 1 with reported IC50 of 1.07 μM.
  • High purity suitable for research use (reported ≥99% / 99.81%).
  • Available in solid pack sizes including 100 MG.
  • Stable as powder when stored at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Used for apoptosis and cancer biology studies in cell models.
Catalog No. 50-002-28871 Supplier Medchemexpress LLC Supplier No. HY115445100MG
LCS-1 is a small-molecule inhibitor of superoxide dismutase 1 (SOD1) used in research to induce apoptosis and study cancer cell biology. The compound is provided as a solid powder and characterized by CAS 41931-13-9, molecular formula C11H8Cl2N2O, and molecular weight 255.10 g·mol-1.
  • Inhibits superoxide dismutase 1 with reported IC50 of 1.07 μM.
  • High purity suitable for research use (reported ≥99% / 99.81%).
  • Available in solid pack sizes including 100 MG.
  • Stable as powder when stored at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Used for apoptosis and cancer biology studies in cell models.