Learn More
Medchemexpress LLC LCS-1 10mg | 41931-13-9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11544510MG
Encompass_Preferred
LCS-1 is a research-grade small molecule used to inhibit superoxide dismutase 1 (SOD1) for studies of oxidative stress and apoptosis. It is supplied as a solid with an indicated purity and storage recommendations for long-term stability.
  • Potent SOD1 inhibitor with reported low-micromolar activity.
  • Molecular formula C11H8Cl2N2O and molecular weight 255.10.
  • Supplied as a light yellow to light brown solid, suitable for formulation in solvents.
  • Includes recommended storage conditions for powder and solutions to preserve stability.
  • Available in small milligram pack sizes for laboratory research use.
Catalog No. 50-002-29171