Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LCS-1 25mg | 41931-13-9 | 255.10 g/mol | C11H8Cl2N2O | 25 MG
SDP

Catalog No. 5000228184
Encompass_Preferred

LCS-1 is a small-molecule inhibitor of superoxide dismutase 1 (SOD1) supplied for preclinical research. It inhibits SOD1 enzymatic activity (reported IC50 = 1.07 μM) and induces apoptosis in multiple cancer cell lines. Provided as a powder for in vitro and in vivo formulation.

  • Molecular formula C11H8Cl2N2O and molecular weight 255.10 g/mol.
  • Potent SOD1 inhibitor; reported IC50 = 1.07 μM.
  • High reported purity (≈99.81%).
  • Solubility: DMSO 50 mg/mL; in vivo formulation ≥2.5 mg/mL in 10% DMSO/90% corn oil.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months).
  • For research use only; not for human or clinical use.

Catalog No. 50-002-28184 Supplier Medchemexpress LLC Supplier No. HY11544525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LCS-1 is a small-molecule inhibitor of superoxide dismutase 1 (SOD1) supplied for preclinical research. It inhibits SOD1 enzymatic activity (reported IC50 = 1.07 μM) and induces apoptosis in multiple cancer cell lines. Provided as a powder for in vitro and in vivo formulation.

  • Molecular formula C11H8Cl2N2O and molecular weight 255.10 g/mol.
  • Potent SOD1 inhibitor; reported IC50 = 1.07 μM.
  • High reported purity (≈99.81%).
  • Solubility: DMSO 50 mg/mL; in vivo formulation ≥2.5 mg/mL in 10% DMSO/90% corn oil.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months).
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.