Learn More
Medchemexpress LLC LCS-1 50mg | 41931-13-9 | 50 MG
SDP
Catalog No. 5000229533
Encompass_Preferred
LCS-1 is a small-molecule inhibitor of superoxide dismutase 1 (SOD1) supplied for laboratory research use. It is provided as a solid and is commonly used in cellular studies probing oxidative stress and apoptosis.
  • Small-molecule SOD1 inhibitor for research use
  • Molecular formula C11H8Cl2N2O; molecular weight 255.10 g/mol
  • Typical purity approximately 99.8% (HPLC)
  • Offered in milligram pack sizes, including 50 mg
  • Powder storage: -20°C (stable for 3 years) or 4°C (stable for 2 years); in solvent: -80°C (6 months), -20°C (1 month)
  • Common applications include cellular assays studying oxidative stress and apoptosis
Catalog No. 50-002-29533 Supplier Medchemexpress LLC Supplier No. HY11544550MG
LCS-1 is a small-molecule inhibitor of superoxide dismutase 1 (SOD1) supplied for laboratory research use. It is provided as a solid and is commonly used in cellular studies probing oxidative stress and apoptosis.
  • Small-molecule SOD1 inhibitor for research use
  • Molecular formula C11H8Cl2N2O; molecular weight 255.10 g/mol
  • Typical purity approximately 99.8% (HPLC)
  • Offered in milligram pack sizes, including 50 mg
  • Powder storage: -20°C (stable for 3 years) or 4°C (stable for 2 years); in solvent: -80°C (6 months), -20°C (1 month)
  • Common applications include cellular assays studying oxidative stress and apoptosis