Learn More
Medchemexpress LLC HY-115445 5mg , LCS-1 CAS:41931-13-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1154455MG
Encompass_Preferred
HY-115445 5mg LCS-1 CAS: 41931-13-9 LCS-1 is a superoxide dismutase 1 (SOD1) inhibitor. LCS-1 inhibits SOD1 activity with an IC50 value of 1.07 μM. LCS-1 induces the early- and late-stage apoptosis of multiple myeloma (MM.1S) cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0572