Learn More
TARGETMOL CHEMICALS INC LCS3 5MG
SDP
Catalog No. 502579618
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. LCS3 is a reversible and non-competitive synergistic inhibitor of glutathione disulfide reductase (GSR) and thioredoxin reductase 1 (TXNRD1) with IC50s of 3.3 uM and 3.8 uM respectively. LCS3 has antitumour activity induces apoptosis and can be used in the study of lung adenocarcinoma (LUAD). purity: 99%
Catalog No. 50-257-9618 Supplier TARGETMOL CHEMICALS INC Supplier No. T604465MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. LCS3 is a reversible and non-competitive synergistic inhibitor of glutathione disulfide reductase (GSR) and thioredoxin reductase 1 (TXNRD1) with IC50s of 3.3 uM and 3.8 uM respectively. LCS3 has antitumour activity induces apoptosis and can be used in the study of lung adenocarcinoma (LUAD). purity: 99%