Learn More
Biomatik Corporation Lct Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11176
Encompass_Preferred
Lct Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LCT. The Lct Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Lactase-Phlorizin Hydrolase Protein (983-1252Aa). Applications: ELISA, IHC. Target Synonyms: LCT. Accession Number: P09848. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9249