Learn More
Waters Corp LCT Premier/XE (Rotary) Performance Maintenance Kit
Catalog No. 50791595 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Performance Maintenance Kit for the LCT Premier/XE (rotary) Mass Spectrometer contains all the Waters Quality Parts necessary to keep your instrument operating at peak performance on an annual basis. The Waters Performance Maintenance Kit includes the following parts: Source component, ESI probe components, reference probe components and vacuum pump components. The original part number was 201000171. This particular part number does not deliver a chemical kit which is required to complete the performance testing. The part number for a Waters Performance Maintenance Kit and a chemical kit is 176002060
Catalog No. 50-791-595 Supplier Waters Corp Supplier No. 201000217
Add to Cart
Add to Cart
Request a Quote
The Waters Performance Maintenance Kit for the LCT Premier/XE (rotary) Mass Spectrometer contains all the Waters Quality Parts necessary to keep your instrument operating at peak performance on an annual basis. The Waters Performance Maintenance Kit includes the following parts: Source component, ESI probe components, reference probe components and vacuum pump components. The original part number was 201000171. This particular part number does not deliver a chemical kit which is required to complete the performance testing. The part number for a Waters Performance Maintenance Kit and a chemical kit is 176002060