Learn More
Waters Corp WATERS CORPORATION Standards Test Kit, LCT Premier/XE, 17 Standards, for Qualification and Calibration
Catalog No. 50201683 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This LCT Premier/XE Sample Kit includes 17 standards to be used for the qualification and calibration of the LCT Premier/XE instrument.
  • Includes 17 standards for qualification and calibration.
  • Designed for LCT Premier and LCT Premier XE instruments.
  • Utilizes reversed phase separation mode.
  • Employs MS detection method.
  • Available in liquid and solid formulations.
  • Store at 2°C to 8°C.
  • Has a shelf life of 12 months.
  • Shipped under ambient conditions.
  • Reference type product.
  • Suitable for system performance and calibration applications.
  • One package contains one unit.
Catalog No. 50-201-683 Supplier Waters Corp Supplier No. 700004027
This LCT Premier/XE Sample Kit includes 17 standards to be used for the qualification and calibration of the LCT Premier/XE instrument.
  • Includes 17 standards for qualification and calibration.
  • Designed for LCT Premier and LCT Premier XE instruments.
  • Utilizes reversed phase separation mode.
  • Employs MS detection method.
  • Available in liquid and solid formulations.
  • Store at 2°C to 8°C.
  • Has a shelf life of 12 months.
  • Shipped under ambient conditions.
  • Reference type product.
  • Suitable for system performance and calibration applications.
  • One package contains one unit.