Learn More
Ted Pella Inc LCY CARBN 300MSH CPR 63UM 25PK
SDP
Catalog No. NC2250199
Encompass
Lacey Carbon, 300 mesh, Copper, approx. grid hole size: 63 um
This item is not returnable. View return policy
Catalog No. NC2250199 Supplier Ted Pella Inc Supplier No. 01895
Lacey Carbon, 300 mesh, Copper, approx. grid hole size: 63 um