Learn More
Electron Microscopy Sciences EMS Lacey Formvar with Carbon Film on 400 mesh Nickel Grid, 150 µm Holes
SDP
Catalog No. 501902775
Encompass_Preferred
This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. Average opening sizes are 50, 100 and 150 microns. This type of pattern provides support but does not interfere when observing specimen section
Grid: Nickel
Holes: 150 µm
Catalog No. 50-190-2775 Supplier Electron Microscopy Sciences Supplier No. LFC400NI150
This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. Average opening sizes are 50, 100 and 150 microns. This type of pattern provides support but does not interfere when observing specimen section
Grid: Nickel
Holes: 150 µm