Learn More
Cayman Chemical LCysteInglutathIon dIsulf 10mg
SDP
Catalog No. 502966449
Encompass_Preferred
A glutathione derivative endogenous to mammalian cells that is comprised of the oxidized form of free glutathione tripeptide linked via a disulfide bond to L-cysteine; protects mice against acetaminophen-induced hepatotoxicity
Catalog No. 50-296-6449 Supplier Cayman Chemical Supplier No. 1758210 MG
A glutathione derivative endogenous to mammalian cells that is comprised of the oxidized form of free glutathione tripeptide linked via a disulfide bond to L-cysteine; protects mice against acetaminophen-induced hepatotoxicity