Learn More
Cayman Chemical L-Cysteinesulfinic Acid (hydrate), Purity: ≥95%, CAS: 207121-48-0, 50 mg
SDP
Catalog No. 502989107
Encompass_Preferred
An excitatory amino acid and mGluR agonist; increases intracellular inositol phosphate levels in CHO cells expressing mGluR1, mGluR5, or mGluR8 (EC50s = 120, 30, and 110 μM, respectively) and inhibits forskolin-induced cAMP production in CHO cells expressing mGluR2 or mGluR6 and hamster kidney cells expressing mGluR4 (EC50s = 100, 100, and 2,000 μM, respectively); selectively binds to mGluR1α (Ki = 3,510 nM) over adrenergic, dopamine, histamine, muscarinic, nicotinic, or serotonin receptors (Kis = >10,000 nM for all); decreases mean arterial blood pressure and heart rate in rats when microinjected into the NTS at 1 mM); can be formed via oxidation of L-cysteine by ROS
Catalog No. 50-298-9107 Supplier Cayman Chemical Supplier No. 2959750 MG
An excitatory amino acid and mGluR agonist; increases intracellular inositol phosphate levels in CHO cells expressing mGluR1, mGluR5, or mGluR8 (EC50s = 120, 30, and 110 μM, respectively) and inhibits forskolin-induced cAMP production in CHO cells expressing mGluR2 or mGluR6 and hamster kidney cells expressing mGluR4 (EC50s = 100, 100, and 2,000 μM, respectively); selectively binds to mGluR1α (Ki = 3,510 nM) over adrenergic, dopamine, histamine, muscarinic, nicotinic, or serotonin receptors (Kis = >10,000 nM for all); decreases mean arterial blood pressure and heart rate in rats when microinjected into the NTS at 1 mM); can be formed via oxidation of L-cysteine by ROS