Learn More
Teledyne ISCO, Inc Ld Class Dual Piston Pump With Belt Drive And Shaped Cams; 0.0 - 100.0 Ml/Min.; 1,000 Psi
Catalog No. 502119944 Shop All Teledyne ISCO Inc. Products
Encompass
LD Class Dual Piston Pump with Belt Drive and Shaped Cams; 0.0 - 100.0 mL/min.; 1,000 psi; Stainless Steel Fluid Path; Automatic Piston Wash; with Pulse Dampener, Pressure & Leak Sensors; Standard Seal (stainless steel energizer); Fitting package for 1/8"""" and 1/16"""" tubing
Catalog No. 50-211-9944 Supplier Teledyne ISCO, Inc Supplier No. LD100SRX
Add to Cart
Add to Cart
Request a Quote
LD Class Dual Piston Pump with Belt Drive and Shaped Cams; 0.0 - 100.0 mL/min.; 1,000 psi; Stainless Steel Fluid Path; Automatic Piston Wash; with Pulse Dampener, Pressure & Leak Sensors; Standard Seal (stainless steel energizer); Fitting package for 1/8"""" and 1/16"""" tubing