Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC American Formula All-Purpose Cleaner, Liquid, Floral Scent, 1 gal

Catalog No. 502839834
Encompass

This all-purpose cleaner utilizes advanced pH neutral floor cleaner technology to effectively emulsify grease, remove dirt, and tackle surface soils in high-traffic areas. Its quick-drying, no-rinse formula ensures no residue or haze is left behind, enhancing floor appearance and extending the life of high-gloss finishes.

  • Liquid form
  • Floral scent
  • Container size of 1 gal
  • pH neutral, ranging from 7.0 to 8.0
  • Dilution ratio of 1 to 4 oz per gallon
  • Red appearance
  • Quick drying and leaves no residue
  • No rinse formula saves time and labor
  • Effectively emulsifies grease and removes dirt

Catalog No. 50-283-9834 Supplier MSC Supplier No. 93843522
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This all-purpose cleaner utilizes advanced pH neutral floor cleaner technology to effectively emulsify grease, remove dirt, and tackle surface soils in high-traffic areas. Its quick-drying, no-rinse formula ensures no residue or haze is left behind, enhancing floor appearance and extending the life of high-gloss finishes.

  • Liquid form
  • Floral scent
  • Container size of 1 gal
  • pH neutral, ranging from 7.0 to 8.0
  • Dilution ratio of 1 to 4 oz per gallon
  • Red appearance
  • Quick drying and leaves no residue
  • No rinse formula saves time and labor
  • Effectively emulsifies grease and removes dirt

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.