Learn More
GL Sciences Inc LD249-B Helium Gas Leak Detector
Catalog No. 5000807370
Encompass
Portable helium gas leak detector based on thermal conductivity detection. Designed for reliable leak checking in laboratory and industrial gas systems. This model is compatible with high magnetic field environments such as MRI systems, allowing safe use where standard detectors may be affected. Suitable for maintenance service troubleshooting and gas line inspection. Provides fast response and easy operation for both routine and specialized applications.
Catalog No. 50-008-07370 Supplier GL Sciences Inc Supplier No. 270219360
Portable helium gas leak detector based on thermal conductivity detection. Designed for reliable leak checking in laboratory and industrial gas systems. This model is compatible with high magnetic field environments such as MRI systems, allowing safe use where standard detectors may be affected. Suitable for maintenance service troubleshooting and gas line inspection. Provides fast response and easy operation for both routine and specialized applications.