Learn More
Mitegen LD35 35L Liquid Dewar By IC Biomedical for Liquid Nitrogen Storage and Dispensing
SDP
Catalog No. 5000495002
Encompass
The IC Biomedical LD35 liquid dewar is a cryogenic vessel designed for storing and dispensing liquid nitrogen. With a 35-liter capacity, this dewar features a rugged construction with a ribbed high-strength aluminum body, a magniformed necktube design, and a more durable paint system. High-performance modern construction and advanced insulation materials ensure high thermal efficiency with a low evaporation rate, providing a static holding time of up to 152 days. It is not intended for sample storage; for that purpose, please see the ICB series SKUs TW-ICB20, TW-ICB24, TW-ICB34, TW-ICB47-10, TW-ICB35-6, TW-ICB35-10 or TW-ICB47-6. The LD35 is lightweight and includes dual handles that allow for easy pouring. It is an ideal solution for labs needing to store and dispense liquid nitrogen.
Catalog No. 50-004-95002 Supplier Mitegen Supplier No. TWLD35
The IC Biomedical LD35 liquid dewar is a cryogenic vessel designed for storing and dispensing liquid nitrogen. With a 35-liter capacity, this dewar features a rugged construction with a ribbed high-strength aluminum body, a magniformed necktube design, and a more durable paint system. High-performance modern construction and advanced insulation materials ensure high thermal efficiency with a low evaporation rate, providing a static holding time of up to 152 days. It is not intended for sample storage; for that purpose, please see the ICB series SKUs TW-ICB20, TW-ICB24, TW-ICB34, TW-ICB47-10, TW-ICB35-6, TW-ICB35-10 or TW-ICB47-6. The LD35 is lightweight and includes dual handles that allow for easy pouring. It is an ideal solution for labs needing to store and dispense liquid nitrogen.