Learn More
Labrepco Inc LD35 SERIES DEWAR
SDP
Catalog No. 509020781
Encompass_Preferred
Dewar, Cryogenic; Labrepco; LD35 series; Ribbed high strength aluminum body, Magniformed necktube design; Capacity: 35L; Weight (empty): 35.1 lb. (16.0kg); Weight (Full): 97.4 lb. (44.3kg); Overall H x dia.: 26.3 x 18.8 in. (668 x 475mm)
Catalog No. 50-902-0781 Supplier Labrepco Inc Supplier No. 35LDB
Dewar, Cryogenic; Labrepco; LD35 series; Ribbed high strength aluminum body, Magniformed necktube design; Capacity: 35L; Weight (empty): 35.1 lb. (16.0kg); Weight (Full): 97.4 lb. (44.3kg); Overall H x dia.: 26.3 x 18.8 in. (668 x 475mm)