Learn More
Labrepco Inc IC Biomedical LD4 Aluminum Cryogenic Dewar, 4 L
SDP
Catalog No. 509020770
Encompass_Preferred
Dewar, Cryogenic; Labrepco; LD4 series; Ribbed high strength aluminum body, Magniformed necktube design; Capacity: 4L; Weight (empty): 6.6 lb. (3.0kg); Weight (Full): 13.7 lb. (6.2kg); Overall H x dia.: 17.0 x 7.6 in. (432 x 193mm)
Catalog No. 50-902-0770 Supplier Labrepco Inc Supplier No. 4LDB
Dewar, Cryogenic; Labrepco; LD4 series; Ribbed high strength aluminum body, Magniformed necktube design; Capacity: 4L; Weight (empty): 6.6 lb. (3.0kg); Weight (Full): 13.7 lb. (6.2kg); Overall H x dia.: 17.0 x 7.6 in. (432 x 193mm)