Learn More
Labrepco Inc LD5 SERIES DEWARS
SDP
Catalog No. 509020772
Encompass_Preferred
Dewar, Cryogenic; Labrepco; LD5 series; Ribbed high strength aluminum body, Magniformed necktube design; Capacity: 5L
Catalog No. 50-902-0772 Supplier Labrepco Inc Supplier No. 5LD
Dewar, Cryogenic; Labrepco; LD5 series; Ribbed high strength aluminum body, Magniformed necktube design; Capacity: 5L