Learn More
Mitegen LD50 50L Liquid Dewar By IC Biomedical for Liquid Nitrogen Storage and Dispensing
SDP
Catalog No. 5000495021
Encompass
The IC Biomedical LD50 liquid dewar is a cryogenic vessel designed for storing and dispensing liquid nitrogen with a large 50-liter capacity. The LD50 features a high-strength aluminum body with a rugged and durable paint system. Its superior vacuum performance, combined with super insulation, ensures maximum holding times and a low evaporation rate, providing a static holding time of up to 122 days. The dewar is designed for easy operation with a lightweight snap-on cap and necktube that provides a tight closure and easy access, along with convenient large handles for easy pouring. This dewar is an ideal solution for laboratories that require a large capacity for storing and dispensing liquid nitrogen. It is not intended for sample storage; for that purpose, please see the ICB series SKUsTW-ICB20, TW-ICB24, TW-ICB34, TW-ICB47-10, TW-ICB35-6, TW-ICB35-10 or TW-ICB47-6.
Catalog No. 50-004-95021 Supplier Mitegen Supplier No. TWLD50
The IC Biomedical LD50 liquid dewar is a cryogenic vessel designed for storing and dispensing liquid nitrogen with a large 50-liter capacity. The LD50 features a high-strength aluminum body with a rugged and durable paint system. Its superior vacuum performance, combined with super insulation, ensures maximum holding times and a low evaporation rate, providing a static holding time of up to 122 days. The dewar is designed for easy operation with a lightweight snap-on cap and necktube that provides a tight closure and easy access, along with convenient large handles for easy pouring. This dewar is an ideal solution for laboratories that require a large capacity for storing and dispensing liquid nitrogen. It is not intended for sample storage; for that purpose, please see the ICB series SKUsTW-ICB20, TW-ICB24, TW-ICB34, TW-ICB47-10, TW-ICB35-6, TW-ICB35-10 or TW-ICB47-6.