Learn More
Medchemexpress LLC LD78-beta/CCL3L1 Hu 100ug
SDP
Catalog No. 5000182869
Encompass_Preferred
LD78-beta/CCL3L1 Protein Human is a potent ligand for the HIV-1 co-receptor CCR5 which is essential for viral entry into human host cells
Catalog No. 50-001-82869 Supplier Medchemexpress LLC Supplier No. HYP7231100UG
Add to Cart
Add to Cart
LD78-beta/CCL3L1 Protein Human is a potent ligand for the HIV-1 co-receptor CCR5 which is essential for viral entry into human host cells