Learn More
Med Vet International LDA Toggles Suture Button, 4in x 15/8in x 1/16in, J0191B, 6 Pack
Catalog No. 5000582241
Encompass_Preferred
This suture button has several different applications in bovine surgery the most common being LDA’s. It is a flat, pliable oval oval-shaped disc with four round, contoured holes. It helps reduce suture tension by distributing the pressure over a wider surface area. Suggested uses: - LDA toggle button repair - Uterine/vaginal prolapse repair - Umbilical hernia in calves - Measures 4" Long x 15/8" Wide x 1/16" Thick - 6 buttons/pack - Neon green color for easy observation - Complete directions included
Catalog No. 50-005-82241 Supplier Med Vet International Supplier No. JOR191B
This suture button has several different applications in bovine surgery the most common being LDA’s. It is a flat, pliable oval oval-shaped disc with four round, contoured holes. It helps reduce suture tension by distributing the pressure over a wider surface area. Suggested uses: - LDA toggle button repair - Uterine/vaginal prolapse repair - Umbilical hernia in calves - Measures 4" Long x 15/8" Wide x 1/16" Thick - 6 buttons/pack - Neon green color for easy observation - Complete directions included