Learn More
Aviva Systems Biology Corp Ldb2 Antibody - N-terminal region (ARP38020_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38020_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ldb2. It was validated on Western Blot. . Alternate names: AI035351, AW146358, CLIM1, CLP-36, Ldb3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-707