Learn More
Aviva Systems Biology Corp LDB3 Antibody - N-terminal region (ARP41588_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41588_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against LDB3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LDB3. Alternate names: CYPHER, KIAA01613, KIAA0613, ORACLE, PDLIM6, ZASP, CMD1C, LVNC3, LDB3Z1, LDB3Z4ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-57917