Learn More
Selleck Chemical LLC LDC000067 10mg 1073485-20-7 LDC067
SDP
Catalog No. 501364814
Encompass_Preferred
LDC000067 (LDC067) is a highly selective CDK9 inhibitor with IC50 of 44 nM, 55/125/210/ >227/ >227-fold selectivity over CDK2/1/4/6/7. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4814 Supplier Selleck Chemical LLC Supplier No. S746110MG
LDC000067 (LDC067) is a highly selective CDK9 inhibitor with IC50 of 44 nM, 55/125/210/ >227/ >227-fold selectivity over CDK2/1/4/6/7. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.