Learn More
Apexbio Technology LLC LDC1267 1361030-48-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063898
Encompass_Preferred
LDC1267 (CAS 1361030-48-9) is a selective inhibitor of the TAM receptor tyrosine kinases Mer Tyro3 and Axl demonstrating IC50 values of less than 5 nM 8 nM and 29 nM respectively By inhibiting TAM signaling LDC1267 abolishes Gas6-mediated suppression in NKG2D-activated natural killer (NK) cells thus enhancing NK-mediated cytotoxicity Preclinical studies show reduced metastasis of melanoma and 4T1 breast tumor cells upon LDC1267 administration in mouse models an effect reliant on NK cell function LDC1267 provides a research tool for investigating TAM-regulated immune modulation in cancer models
Catalog No. 50-000-63898 Supplier Apexbio Technology LLC Supplier No. B48935.1
LDC1267 (CAS 1361030-48-9) is a selective inhibitor of the TAM receptor tyrosine kinases Mer Tyro3 and Axl demonstrating IC50 values of less than 5 nM 8 nM and 29 nM respectively By inhibiting TAM signaling LDC1267 abolishes Gas6-mediated suppression in NKG2D-activated natural killer (NK) cells thus enhancing NK-mediated cytotoxicity Preclinical studies show reduced metastasis of melanoma and 4T1 breast tumor cells upon LDC1267 administration in mouse models an effect reliant on NK cell function LDC1267 provides a research tool for investigating TAM-regulated immune modulation in cancer models