Learn More
Apexbio Technology LLC LDC1267, 5mg. Cas: 1361030-48-9 MFCD: MFCD28385861. TAM kinase inhibitor,highly selective
SDP
Supplier:  Apexbio Technology LLC B48935
Encompass_Preferred
LDC1267 is a highly selective TAM kinase inhibitor with IC50 of < 5 nM, 8 nM, and 29 nM for Mer, Tyro3, and Axl, respectively. LDC1267 preferentially inhibits Tyro3, Axl and Mer at low nano-molarity by tracer-based binding assays. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5363