Learn More
Medchemexpress LLC LDC4297 (hydrochloride) | 2319747-14-1 | 98.2% | 25 MG
SDP
Catalog No. 5000338873
Encompass_Preferred
LDC4297 hydrochloride is a selective inhibitor of CDK7, effectively inhibiting human cytomegalovirus (HCMV) replication. It exhibits broad antiviral activities against various virus families including Herpesviridae, Adenoviridae, Poxviridae, Retroviridae, and Orthomyxoviridae. This compound is suitable for infection research.
  • Selective inhibitor of CDK7
  • Inhibits human cytomegalovirus (HCMV) replication
  • Demonstrates broad antiviral activities against multiple virus families
  • Useful for infection research
  • Solid form storage: 4°C, sealed, away from moisture and light
  • In-solvent storage: -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light
Catalog No. 50-003-38873 Supplier Medchemexpress LLC Supplier No. 25MGHY12653A
LDC4297 hydrochloride is a selective inhibitor of CDK7, effectively inhibiting human cytomegalovirus (HCMV) replication. It exhibits broad antiviral activities against various virus families including Herpesviridae, Adenoviridae, Poxviridae, Retroviridae, and Orthomyxoviridae. This compound is suitable for infection research.
  • Selective inhibitor of CDK7
  • Inhibits human cytomegalovirus (HCMV) replication
  • Demonstrates broad antiviral activities against multiple virus families
  • Useful for infection research
  • Solid form storage: 4°C, sealed, away from moisture and light
  • In-solvent storage: -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light