Learn More
Medchemexpress LLC LDC4297 hydrochloride | 2319747-14-1 | 98.2% | 50 MG
SDP
Catalog No. 5000342239
Encompass_Preferred
LDC4297 hydrochloride is a selective inhibitor of CDK7 with an IC50 value of 0.13 nM. It inhibits human cytomegalovirus (HCMV) replication with an EC50 value of 24.5 nM. This compound demonstrates broad antiviral activities against Herpesviridae, Adenoviridae, Poxviridae, Retroviridae, and Orthomyxoviridae, with EC50 values ranging from 0.02-1.21 μM. It is suitable for research related to infection.
  • Selective inhibitor of CDK7
  • Inhibits human cytomegalovirus (HCMV) replication
  • Broad antiviral activities against various viruses
  • Suitable for infection research
  • Exhibits anti-proliferative activity in human fibroblasts
  • Demonstrates good oral bioavailability and half-life in vivo
Catalog No. 50-003-42239 Supplier Medchemexpress LLC Supplier No. HY12653A50MG
LDC4297 hydrochloride is a selective inhibitor of CDK7 with an IC50 value of 0.13 nM. It inhibits human cytomegalovirus (HCMV) replication with an EC50 value of 24.5 nM. This compound demonstrates broad antiviral activities against Herpesviridae, Adenoviridae, Poxviridae, Retroviridae, and Orthomyxoviridae, with EC50 values ranging from 0.02-1.21 μM. It is suitable for research related to infection.
  • Selective inhibitor of CDK7
  • Inhibits human cytomegalovirus (HCMV) replication
  • Broad antiviral activities against various viruses
  • Suitable for infection research
  • Exhibits anti-proliferative activity in human fibroblasts
  • Demonstrates good oral bioavailability and half-life in vivo