Learn More
Medchemexpress LLC LDC4297 hydrochloride | 2319747-14-1 | 98.23% | 100 MG
SDP
Catalog No. 5000339155
Encompass_Preferred
LDC4297 hydrochloride is a selective inhibitor of CDK7 with an IC50 value of 0.13 nM. It inhibits human cytomegalovirus (HCMV) replication with an EC50 value of 24.5 nM. This compound demonstrates broad antiviral activities against various viruses, including Herpesviridae, Adenoviridae, Poxviridae, Retroviridae, and Orthomyxoviridae, with EC50 values ranging from 0.02-1.21 μM. LDC4297 hydrochloride is suitable for infection research.
  • Selective CDK7 inhibitor (IC50: 0.13 nM)
  • Inhibits human cytomegalovirus (HCMV) replication (EC50: 24.5 nM)
  • Exhibits broad antiviral activities against various viruses (EC50: 0.02-1.21 μM)
  • Useful for infection research
Catalog No. 50-003-39155 Supplier Medchemexpress LLC Supplier No. HY12653A100MG
LDC4297 hydrochloride is a selective inhibitor of CDK7 with an IC50 value of 0.13 nM. It inhibits human cytomegalovirus (HCMV) replication with an EC50 value of 24.5 nM. This compound demonstrates broad antiviral activities against various viruses, including Herpesviridae, Adenoviridae, Poxviridae, Retroviridae, and Orthomyxoviridae, with EC50 values ranging from 0.02-1.21 μM. LDC4297 hydrochloride is suitable for infection research.
  • Selective CDK7 inhibitor (IC50: 0.13 nM)
  • Inhibits human cytomegalovirus (HCMV) replication (EC50: 24.5 nM)
  • Exhibits broad antiviral activities against various viruses (EC50: 0.02-1.21 μM)
  • Useful for infection research