Learn More
Medchemexpress LLC LDC4297 hydrochloride | 2319747-14-1 | MFCD34676707 | 98.2% | 468.98 | C23H29ClN8O | 5 MG
SDP
Catalog No. 5000264070
Encompass_Preferred
LDC4297 hydrochloride is a small-molecule research reagent that selectively inhibits cyclin-dependent kinase 7 (CDK7). Supplied as the hydrochloride salt for laboratory use only, it is intended for cell-signaling and antiviral research and is available in both solid and solution formats.
  • Selective CDK7 inhibition for target-specific studies.
  • Suitable for in vitro cell-based assays and mechanistic studies.
  • Available in multiple sizes including small solid quantities and DMSO solution.
  • High purity suitable for research applications.
  • Supplied as the hydrochloride salt for improved stability and solubility.
Catalog No. 50-002-64070 Supplier Medchemexpress LLC Supplier No. HY12653A5MG
LDC4297 hydrochloride is a small-molecule research reagent that selectively inhibits cyclin-dependent kinase 7 (CDK7). Supplied as the hydrochloride salt for laboratory use only, it is intended for cell-signaling and antiviral research and is available in both solid and solution formats.
  • Selective CDK7 inhibition for target-specific studies.
  • Suitable for in vitro cell-based assays and mechanistic studies.
  • Available in multiple sizes including small solid quantities and DMSO solution.
  • High purity suitable for research applications.
  • Supplied as the hydrochloride salt for improved stability and solubility.