Learn More
Medchemexpress LLC HY-111674 10mg Medchemexpress, LDC7559 CAS: Purity:>98%
SDP
Catalog No. 501968943
Encompass_Preferred
Medchemexpress, HY-111674 10mg LDC7559 CAS: LDC7559 is a gasdermin D (GSDMD) inhibitor via blocking neutrophil extracellular trap (NET) in the late stages . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8943 Supplier Medchemexpress LLC Supplier No. HY11167410MG
Medchemexpress, HY-111674 10mg LDC7559 CAS: LDC7559 is a gasdermin D (GSDMD) inhibitor via blocking neutrophil extracellular trap (NET) in the late stages . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.