Learn More
Medchemexpress LLC HY-111674 5mg Medchemexpress, LDC7559 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1116745MG
Encompass_Preferred
Medchemexpress, HY-111674 5mg LDC7559 CAS: LDC7559 is a gasdermin D (GSDMD) inhibitor via blocking neutrophil extracellular trap (NET) in the late stages [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2384