Learn More
Medchemexpress LLC HY-16582A 100mg , LDE225 NVP-LDE 225;Erismodegib CAS:956697-53-3 Purity:98%
SDP
Catalog No. 501493066
Encompass_Preferred
Medchemexpress, HY-16582A 100mg LDE225 NVP-LDE 225;Erismodegib CAS:956697-53-3 Formula:C26H26F3N3O3 IC50: 1.3 nM (mSmo), 2.5 nM (hSmo) Purity:98% LDE225 is a potent and selective Smo antagonist with IC 50 of 1.3 nM and 2.5 nM for mouse and human Smo in binding assay, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3066 Supplier Medchemexpress LLC Supplier No. HY16582A100MG
Medchemexpress, HY-16582A 100mg LDE225 NVP-LDE 225;Erismodegib CAS:956697-53-3 Formula:C26H26F3N3O3 IC50: 1.3 nM (mSmo), 2.5 nM (hSmo) Purity:98% LDE225 is a potent and selective Smo antagonist with IC 50 of 1.3 nM and 2.5 nM for mouse and human Smo in binding assay, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.