Learn More
Apexbio Technology LLC LDE225 Diphosphate 1218778-77-8 5mg
SDP
Catalog No. 5000094264
Encompass_Preferred
LDE225 Diphosphate (CAS 1218778-77-8) is a highly potent and selective antagonist of the Smoothened (Smo) receptor a seven-transmembrane protein involved in the Hedgehog (Hh) signaling pathway LDE225 inhibits Smo with IC50 values of 1 3 nM for murine and 2 5 nM for human Smo resulting in suppression of Hh signaling This pathway is implicated in cellular differentiation proliferation and oncogenesis In vitro LDE225 selectively binds Smo inhibiting Hh-dependent tumor cell growth In vivo it demonstrates dose-dependent antitumor efficacy in medulloblastoma xenograft mouse models LDE225 continues to be explored as a research tool and therapeutic candidate in Hh pathway-driven malignancies
Catalog No. 50-000-94264 Supplier Apexbio Technology LLC Supplier No. A35435
LDE225 Diphosphate (CAS 1218778-77-8) is a highly potent and selective antagonist of the Smoothened (Smo) receptor a seven-transmembrane protein involved in the Hedgehog (Hh) signaling pathway LDE225 inhibits Smo with IC50 values of 1 3 nM for murine and 2 5 nM for human Smo resulting in suppression of Hh signaling This pathway is implicated in cellular differentiation proliferation and oncogenesis In vitro LDE225 selectively binds Smo inhibiting Hh-dependent tumor cell growth In vivo it demonstrates dose-dependent antitumor efficacy in medulloblastoma xenograft mouse models LDE225 continues to be explored as a research tool and therapeutic candidate in Hh pathway-driven malignancies