Learn More
Selleck Chemical LLC Sonidegib (NVP-LDE225) 5mg 956697-53-3 Erismodegib
SDP
Supplier:  Selleck Chemical LLC S21515MG
Encompass_Preferred
Sonidegib (Erismodegib, NVP-LDE225) is a Smoothened (Smo) antagonist, inhibiting Hedgehog (Hh) signaling with IC50 of 1.3 nM (mouse) and 2.5 nM (human) in cell-free assays, respectively. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0747234