Learn More
Cayman Chemical LDE225phosphate 10mg
SDP
Catalog No. 502971151
Encompass_Preferred
A Smo antagonist (IC50s = 1.3 and 2.5 nM for mouse and human Smo, respectively); inhibits Gli1 mRNA expression and prevents tumor growth in an orthotopic Ptch+/- p53-/- medulloblastoma mouse allograft model at 20 mg/kg per day
Catalog No. 50-297-1151 Supplier Cayman Chemical Supplier No. 1626310 MG
A Smo antagonist (IC50s = 1.3 and 2.5 nM for mouse and human Smo, respectively); inhibits Gli1 mRNA expression and prevents tumor growth in an orthotopic Ptch+/- p53-/- medulloblastoma mouse allograft model at 20 mg/kg per day