Learn More
ABclonal Technology LDH-B
SDP
Supplier:  ABclonal Technology A2758420UL
Encompass_Preferred
This gene encodes the B subunit of lactate dehydrogenase enzyme which catalyzes the interconversion of pyruvate and lactate with concomitant interconversion of NADH and NAD in a post-glycolysis process Alternatively spliced transcript variants have been found for this gene Recent studies have shown that a C-terminally extended isoform is produced by use of an alternative in-frame translation termination codon via a stop codon readthrough mechanism and that this isoform is localized in the peroxisomes Mutations in this gene are associated with lactate dehydrogenase B deficiency Pseudogenes have been identified on chromosomes X 5 and 13
Catalog No. 50-000-13299