Learn More
Roche Diagnostics LDH Bio HTTest kit for the Cedex Bio HT Analyzer
Catalog No. 50589416 Shop All Roche Diagnostics Products
Encompass
LDH Bio HT. Test kit for the Cedex Bio HT Analyzer for use in quality control / manufacturing process only. 300 tests. Method: Enzymatic photometric assay using oxidation of lactate to pyruvate (acc. IFCC). Recovery: 10%
This item is not returnable. View return policy
Catalog No. 50-589-416 Supplier Roche Diagnostics Supplier No. 06608493001
LDH Bio HT. Test kit for the Cedex Bio HT Analyzer for use in quality control / manufacturing process only. 300 tests. Method: Enzymatic photometric assay using oxidation of lactate to pyruvate (acc. IFCC). Recovery: 10%