Learn More
TRIBIOSCIENCE INC Lactate Dehydrogenase (LDH) Cytotoxicity Assay Kit, 500 Tests
SDP
Catalog No. NC2886686
Encompass_Preferred
LDH Cytotoxicity Colorimetric Assay Kit provides the easiest and most accurate approach to measure LDH activity and cytotoxicity of a variety of compounds. The generated signal at OD 460 nm is proportional to the LDH levels of lysed cells.
Catalog No. NC2886686 Supplier TRIBIOSCIENCE INC Supplier No. TBS2002
LDH Cytotoxicity Colorimetric Assay Kit provides the easiest and most accurate approach to measure LDH activity and cytotoxicity of a variety of compounds. The generated signal at OD 460 nm is proportional to the LDH levels of lysed cells.