Learn More
Cell Biolabs Inc CytoSelect™ LDH Cytotoxicity Assay Kit, 960 assays
Catalog No. 50672680
Encompass_Preferred
The CytoSelect™ LDH Cytotoxicity Assay Kit provides a colorimetric format for measuring cell cytotoxicity. Cells can be plated and then treated with compounds or agents that affect cell viability. Upon cell death, lactate dehydrogenase (LDH), a soluble enzyme found in the cytoplasm, is released into the growth media. In the presence of lactate substrate, LDH converts lactate to pyruvate and generates nicotinamide adenine dinucleotide (NADH). The WST-1 molecule is converted to the orange formazan form. An increase in cell cytotoxicity is accompanied by increased LDH release and increased colorimetric signal. The assay principles are basic and can be applied to most eukaryotic cell lines, including adherent and non-adherent cells and certain tissues, depending on LDH expression levels. The LDH Cytotoxicity Reagent can be used to detect cytotoxicity in mammalian cells. The kit contains sufficient reagents for the evaluation of 960 assays in 96-well plates.
Catalog No. 50-672-680 Supplier Cell Biolabs Inc Supplier No. CBA241
The CytoSelect™ LDH Cytotoxicity Assay Kit provides a colorimetric format for measuring cell cytotoxicity. Cells can be plated and then treated with compounds or agents that affect cell viability. Upon cell death, lactate dehydrogenase (LDH), a soluble enzyme found in the cytoplasm, is released into the growth media. In the presence of lactate substrate, LDH converts lactate to pyruvate and generates nicotinamide adenine dinucleotide (NADH). The WST-1 molecule is converted to the orange formazan form. An increase in cell cytotoxicity is accompanied by increased LDH release and increased colorimetric signal. The assay principles are basic and can be applied to most eukaryotic cell lines, including adherent and non-adherent cells and certain tissues, depending on LDH expression levels. The LDH Cytotoxicity Reagent can be used to detect cytotoxicity in mammalian cells. The kit contains sufficient reagents for the evaluation of 960 assays in 96-well plates.