Learn More
Cayman Chemical LDH Cytotoxicity Assay Kit, 480 Well
SDP
Catalog No. 502896364
Encompass_Preferred
For measurement of LDH activity as an indicator of cell death
Catalog No. 50-289-6364 Supplier Cayman Chemical Supplier No. 601170480 WELL
For measurement of LDH activity as an indicator of cell death