Learn More
Medchemexpress LLC HY-111108 5mg Medchemexpress, LDH-IN-1 CAS:1964515-43-2 Purity:>98%
SDP
Catalog No. 501968822
Encompass_Preferred
Medchemexpress, HY-111108 5mg LDH-IN-1 CAS:1964515-43-2 LDH-IN-1 is a novel pyrazole-based inhibitor of human lactate dehydrogenase (LDH) with IC50s of 32 and 27 nM for LDHA and LDHB, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8822 Supplier Medchemexpress LLC Supplier No. HY1111085MG
Medchemexpress, HY-111108 5mg LDH-IN-1 CAS:1964515-43-2 LDH-IN-1 is a novel pyrazole-based inhibitor of human lactate dehydrogenase (LDH) with IC50s of 32 and 27 nM for LDHA and LDHB, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.