Learn More
TRIBIOSCIENCE INC LDH Lactate Dehydrogenase Colorimetric Assay Kit, 500 Tests
SDP
Catalog No. NC2887481
Encompass_Preferred
LDH Colorimetric Assay Kit provides the easiest and most accurate approach to measure LDH activity. The generated signal at OD 460 nm is proportional to the LDH released from the lysed cells.
Catalog No. NC2887481 Supplier TRIBIOSCIENCE INC Supplier No. TBS2012
LDH Colorimetric Assay Kit provides the easiest and most accurate approach to measure LDH activity. The generated signal at OD 460 nm is proportional to the LDH released from the lysed cells.