Learn More
Enzo Life Sciences Lactate Dehydrogenase A monoclonal antibody (AT1A4) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS7310100
Encompass_Preferred
Clone: AT1A4. Isotype: IgG2bκ. Immunogen: Recombinant human LDHA (aa 1-332) purified from E. coli. Source: Purified from ascites. Species reactivity: Human. Applications: ELISA, Flow Cytometry, ICC, IF, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide and 10% glycerol.
Catalog No. 50-201-3272