Learn More
TARGETMOL CHEMICALS INC LDHA-IN-4 10MG
SDP
Catalog No. 502580352
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. LDHA-IN-4 (AZ33) is an LDHA inhibitor. Binding affinity to 6-His-tagged human LDHA (2 to 322 amino acid residue) expressed in Escherichia coli BL21 (DE3) cells by SPR analysis with an active value of 0.093 uM. purity: 99%
Catalog No. 50-258-0352 Supplier TARGETMOL CHEMICALS INC Supplier No. T317510MG
Add to Cart
Add to Cart
Also available in 1 mg 5 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. LDHA-IN-4 (AZ33) is an LDHA inhibitor. Binding affinity to 6-His-tagged human LDHA (2 to 322 amino acid residue) expressed in Escherichia coli BL21 (DE3) cells by SPR analysis with an active value of 0.093 uM. purity: 99%