Learn More
Aviva Systems Biology Corp LDHAL6A Antibody - C-terminal region (ARP63123_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63123_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LDHAL6A. It was validated on Western Blot. . Alternate names: LDH6A, MGC23940ApplicationWB. Species reactivity: Cow, Human, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-887