Learn More
Aviva Systems Biology Corp LDHC Antibody - middle region (ARP53602_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53602_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LDHC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LDHC. Alternate names: LDH3, LDHX, MGC111073, CT32ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61065